Propyl-4-hydroxybenzoate ≥99%

Supplier: BeanTown Chemical

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

135805-1KG 135805-250G 135805-5KG
BT135805-1KGEA 92 USD
BT135805-1KG BT135805-250G BT135805-5KG
Propyl-4-hydroxybenzoate ≥99%
Propyl-4-hydroxybenzoate

CAS: 94-13-3; EC No: 202-307-7; MDL No: MFCD00002354; RTECS: DH2800000 Crystalline; Linear Formula: HOC6H4COOCH2CH2CH3; Molecular Formula: C10H12O3 ; MW: 180.20 Melting Point: 95-98°; Boiling Point: 301°; Flash point: 180°C (356°F) Density (g/mL): 1.287


  • RTECS: DH2800000
  • Crystalline

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 301 °C
Melting Pt: 95…98 °C
Density: 1.287 g/mL
Flash Pt: 180 °C (356 °F)
MDL Number: MFCD00002354
CAS Number: 94-13-3
EINECS: 202-307-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR