Trimethyl phosphate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Trimethylphosphate
135785-2.5KG
135785-500G
BT135785-2.5KGEA
260
USD
BT135785-2.5KG
BT135785-500G
Trimethyl phosphate ≥98%
Trimethyl phosphate
CAS: 512-56-1; EC No: 208-144-8; MDL No: MFCD00008348; RTECS: TC8225000
Liquid; Linear Formula: P(O)(OCH3)3; MW: 140.08
Melting Point: -46°; Boiling Point: 196-198°; Flash point: 150°C (302°F)
Density (g/mL): 1.197; Refractive Index: 1.395
Moisture Sensitive
- RTECS: TC8225000
- Refractive index: 1.395
- Moisture Sensitive
- Liquid
Formula:
C₃H₉O₄P MW: 140.08 g/mol Boiling Pt: 196…198 °C Melting Pt: –46 °C Density: 1.197 g/mL Flash Pt: 150 °C (302 °F) |
MDL Number:
MFCD00008348 CAS Number: 512-56-1 EINECS: 208-144-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...