Chromium(III) potassium sulfate dodecahydrate ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
135775-100G
135775-25G
135775-500G
BT135775-100GEA
381
USD
BT135775-100G
BT135775-25G
BT135775-500G
Chromium(III) potassium sulfate dodecahydrate ≥99.9% (trace metals basis)
Chromium(III) potassium sulphate dodecahydrate
CAS: 7788-99-0; EC No: 233-401-6; MDL No: MFCD00149917; RTECS: GB6850000
Crystalline; Linear Formula: CrK(SO4)2·12H2O ; MW: 499.39
Melting Point: 89°
- RTECS: GB6850000
- Crystalline
Formula:
CrK(SO₄)₂·12H₂O Melting Pt: 89 °C |
MDL Number:
MFCD00149917 CAS Number: 7788-99-0 EINECS: 233-401-6 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...