L(-)-Tyrosine ≥99%

Supplier: BeanTown Chemical

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

135745-250G 135745-50G
BT135745-250GEA 121.08 USD
BT135745-250G BT135745-50G
L(-)-Tyrosine ≥99%
L(-)-Tyrosine

CAS: 60-18-4; EC No: 200-460-4; MDL No: MFCD00002606; RTECS: YP2275600 Crystalline/Pieces; Molecular Formula: C9H11NO3; MW: 181.19


  • RTECS: YP2275600
  • Crystalline/Pieces

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
Boiling Pt: 351
Melting Pt: 285
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 13,09907

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR