Hydroquinone monomethyl ether ≥98%
Supplier: BeanTown Chemical
Synonyms:
p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole
135730-1KG
135730-250G
135730-50G
BT135730-1KGEA
131.96
USD
BT135730-1KG
BT135730-250G
BT135730-50G
Hydroquinone monomethyl ether ≥98%
Hydroquinone monomethyl ether
CAS: 150-76-5; EC No: 205-769-8; MDL No: MFCD00002332; RTECS: SL7700000
Crystalline; Molecular Formula: C7H8O2; MW: 124.14
Melting Point: 55-57°; Boiling Point: 243°
- RTECS: SL7700000
- Crystalline
Formula:
C₇H₈O₂ MW: 124.14 g/mol Boiling Pt: 243 Melting Pt: 55…57 °C Flash Pt: 128 |
MDL Number:
MFCD00002332 CAS Number: 150-76-5 EINECS: 205-769-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...