Iminodiacetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,2'-Azanediyldiacetic acid

135705-100G 135705-2.5KG 135705-500G
BT135705-100GEA 26.55 USD
BT135705-100G BT135705-2.5KG BT135705-500G
Iminodiacetic acid ≥98%
Iminodiacetic acid

CAS: 142-73-4; EC No: 205-555-4; MDL No: MFCD00004280; RTECS: AI2975000 Crystalline; Linear Formula: HN(CH2CO2H)2 ; Molecular Formula: C4H7NO4; MW: 133.10 Melting Point: 243° (decomposes)


  • RTECS: AI2975000
  • Crystalline

Formula: C₄H₇NO₄
MW: 133.10 g/mol
Melting Pt: 243 °C (dec.)
MDL Number: MFCD00004280
CAS Number: 142-73-4
EINECS: 205-555-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR