Diethyl acetylmalonate ≥85%, Technical Grade
Supplier: BeanTown Chemical
135700-1G
135700-5G
BT135700-1GEA
29
USD
BT135700-1G
BT135700-5G
Diethyl acetylmalonate ≥85%, Technical Grade
Diethyl acetylmalonate
CAS: 570-08-1; MDL No: MFCD00026876
Liquid; Molecular Formula: C9H14O5; MW: 202.21
Boiling Point: 119-120°/17 mmHg
- Liquid
Formula:
C₉H₁₄O₅ Boiling Pt: 119…120 °C/17 mmHg |
MDL Number:
MFCD00026876 CAS Number: 570-08-1 |
Specification Test Results
Assay | 85% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...