Diphenylacetylene ≥99%

Supplier: BeanTown Chemical

135680-100G 135680-25G 135680-5G
BT135680-100GEA 275.56 USD
BT135680-100G BT135680-25G BT135680-5G
Diphenylacetylene ≥99%
Diphenylacetylene

CAS: 501-65-5; EC No: 207-926-6; MDL No: MFCD00004786 Crystalline; Linear Formula: C6H5C≡CC6H5; Molecular Formula: C14H10; MW: 178.23 Melting Point: 59-61°; Boiling Point: 170°/19 mmHg Density (g/mL): 0.99


  • Crystalline

Formula: C₁₄H₁₀
MW: 178.23 g/mol
Boiling Pt: 170 °C/19 mmHg
Melting Pt: 59…61 °C
Density: 0.99 g/mL
MDL Number: MFCD00004786
CAS Number: 501-65-5
EINECS: 207-926-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR