Rubidium carbonate ≥99.95% (trace metals basis)

Supplier: BeanTown Chemical

135675-1G 135675-25G 135675-5G
BT135675-1GEA 38.73 USD
BT135675-1G BT135675-25G BT135675-5G
Rubidium carbonate ≥99.95% (trace metals basis)
Rubidium carbonate

CAS: 584-09-8; EC No: 209-530-9; MDL No: MFCD00011188; RTECS: FG0650000 Powder; Linear Formula: Rb2CO3; MW: 230.95 Melting Point: 837° Hygroscopic


  • RTECS: FG0650000
  • Hygroscopic
  • Powder

Formula: CO₃Rb₂
Melting Pt: 837 °C
MDL Number: MFCD00011188
CAS Number: 584-09-8
EINECS: 209-530-9

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR