Rhodium(III) chloride hydrate ≥99.99% (trace metals basis)
Supplier: BeanTown Chemical
135670-1G
135670-5G
BT135670-1GEA
496
USD
BT135670-1G
BT135670-5G
Rhodium(III) chloride hydrate ≥99.99% (trace metals basis)
Rhodium(III) chloride hydrate
CAS: 20765-98-4; EC No: 233-165-4; MDL No: MFCD00149839
Crystalline; Linear Formula: RhCl3·xH2O; MW: 209.26 (anhydrous)
Melting Point: 100° (decomposes)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Cl₃Rh·H₂O Melting Pt: 100 °C (dec.) |
MDL Number:
MFCD00149839 CAS Number: 20765-98-4 EINECS: 233-165-4 |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...