Pyridinium-p-toluenesulphonate ≥98%

Supplier: BeanTown Chemical

Synonyms: Pyridin-1-ium 4-methylbenzenesulphonate

135655-100G 135655-25G 135655-500G
BT135655-100GEA 183 USD
BT135655-100G BT135655-25G BT135655-500G
Pyridinium-p-toluenesulphonate ≥98%
Pyridinium-p-toluenesulphonate

CAS: 24057-28-1; EC No: 246-002-7; MDL No: MFCD00013108 Crystalline/Powder; Molecular Formula: C12H13NO3S; MW: 251.31 Melting Point: 117-119° Moisture Sensitive


  • Moisture Sensitive
  • Crystalline/Powder

Formula: C₁₂H₁₃NO₃S
MW: 251.31 g/mol
Melting Pt: 117…119 °C
MDL Number: MFCD00013108
CAS Number: 24057-28-1
EINECS: 246-002-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR