Dipotassium hexachloroosmate

Supplier: BeanTown Chemical

Synonyms: Potassium hexachloroosmate(IV)

135645-1G 135645-250MG 135645-5G
BT135645-1GEA 204 USD
BT135645-1G BT135645-250MG BT135645-5G
Dipotassium hexachloroosmate
Dipotassium hexachloroosmate

CAS: 16871-60-6; EC No: 240-893-6; MDL No: MFCD00011371 Powder; Linear Formula: K2OsCl6; MW: 481.12 Melting Point: 600° (decomposes) Density (g/mL): 3.42 Hygroscopic


  • Hygroscopic
  • Powder

Formula: Cl₆K₂Os
MW: 481.14 g/mol
Melting Pt: 600 °C (dec.)
Density: 3.42 g/mL
MDL Number: MFCD00011371
CAS Number: 16871-60-6
EINECS: 240-893-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR