3,4,5-Trichloropyridazine

Supplier: BeanTown Chemical

135585-1G 135585-5G
BT135585-1GEA 130 USD
BT135585-1G BT135585-5G
3,4,5-Trichloropyridazine
3,4,5-Trichloropyridazine

CAS: 14161-11-6; EC No: 238-006-2 Solid; Molecular Formula: C4HCl3N2; MW: 183.42


  • Solid

Formula: C₄HCl₃N₂
MW: 183.42 g/mol
MDL Number: MFCD00006464
CAS Number: 14161-11-6
EINECS: 238-006-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR