Ruthenium(III) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)

135580-1G 135580-5G
BT135580-1GEA 87 USD
BT135580-1G BT135580-5G
Ruthenium(III) acetylacetonate
Ruthenium(III) acetylacetonate

CAS: 14284-93-6; EC No: 238-193-0; MDL No: MFCD00000030 Crystalline; Linear Formula: Ru(C5H7O2)3; MW: 398.40 Melting Point: 260° (decomposes)


  • Crystalline

Formula: C₁₅H₂₁O₆Ru
MW: 398.4 g/mol
Melting Pt: 260 °C (dec.)
MDL Number: MFCD00000030
CAS Number: 14284-93-6
EINECS: 238-193-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR