Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex in vinyl terminated polydimethylsiloxane

Supplier: BeanTown Chemical

Synonyms: Karstedt Catalyst, Platinum(0) - 1,3-Divinyltetramethyldisiloxane Complex

135575-1G 135575-25G 135575-5G
BT135575-1GEA 82.32 USD
BT135575-1G BT135575-25G BT135575-5G
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex in vinyl terminated polydimethylsiloxane
Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex

CAS: 68478-92-2; MDL No: MFCD00151662 Liquid; Linear Formula: C8H18OPtSi2; MW: 381.48 Boiling Point: 200°; Flash point: 110°C (230°F) Density (g/mL): 0.984 Moisture Sensitive


  • Moisture Sensitive
  • Liquid

Formula: C₈H₁₈OPtSi₂
MW: 381.485 g/mol
Boiling Pt: >200 °C
Density: 0.984 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00151662
CAS Number: 68478-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR