5-Bromo-2-fluoro-4-methylpyridine ≥98%

Supplier: BeanTown Chemical

135550-100G 135550-25G 135550-5G
BT135550-100GEA 556 USD
BT135550-100G BT135550-25G BT135550-5G
5-Bromo-2-fluoro-4-methylpyridine ≥98%
5-Bromo-2-fluoro-4-methylpyridine

CAS: 864830-16-0; MDL No: MFCD09839213 Liquid; Molecular Formula: C6H5BrFN; MW: 190.01


  • Liquid

Formula: C₆H₅BrFN
MDL Number: MFCD09839213
CAS Number: 864830-16-0

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR