Platinum ≥99.95% (trace metals basis), wire Ø 1.0 mm, hard type

Supplier: BeanTown Chemical

Synonyms: Platinium

135540-25CM 135540-5CM
BT135540-25CMEA 969.28 USD
BT135540-25CM BT135540-5CM
Platinum ≥99.95% (trace metals basis), wire Ø 1.0 mm, hard type
Platinum

CAS: 7440-06-4; EC No: 231-116-1; MDL No: MFCD00011179; RTECS: TP2160000 Solid; Molecular Formula: Pt; MW: 195.08 Melting Point: 1772°; Boiling Point: 3827° Density (g/mL): 21.45


  • RTECS: TP2160000
  • Solid

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.45 g/mL
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR