3-Amino-5-cyclobutyl-1H-pyrazole ≥97%
Supplier: BeanTown Chemical
135525-1G
135525-250MG
BT135525-1GEA
276.55
USD
BT135525-1G
BT135525-250MG
3-Amino-5-cyclobutyl-1H-pyrazole ≥97%
3-Amino-5-cyclobutyl-1H-pyrazole
CAS: 326827-21-8; MDL No: MFCD08061045
Liquid; Molecular Formula: C7H11N3; MW: 137.19
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₇H₁₁N₃ |
MDL Number:
MFCD08061045 CAS Number: 326827-21-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...