Bathocuproin ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

135510-1G 135510-5G
BT135510-1GEA 74 USD
BT135510-1G BT135510-5G
Bathocuproin ≥98%
Bathocuproin

CAS: 4733-39-5; EC No: 225-240-5; MDL No: MFCD00004972 Powder; Molecular Formula: C26H20N2; MW: 360.45 Melting Point: 279-283°


  • Powder

Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Melting Pt: 279…283 °C
MDL Number: MFCD00004972
CAS Number: 4733-39-5
EINECS: 225-240-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR