Cerium(IV) oxide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Ceric oxide, Cerium dioxide
135495-100G
135495-25G
135495-500G
BT135495-100GEA
131
USD
BT135495-100G
BT135495-25G
BT135495-500G
Cerium(IV) oxide ≥99.5%
Cerium(IV) oxide
CAS: 1306-38-3; EC No: 215-150-4; MDL No: MFCD00010933; RTECS: FK6310000
15-30nm APS Powder, S.A. 30-50 m2/g; Molecular Formula: CeO2; MW: 172.12
Density (g/mL): 7.13
- RTECS: FK6310000
- 15-30nm APS Powder, S.A. 30-50 m²/g
Formula:
CeO₂ MW: 172.11 g/mol Boiling Pt: 3468 Melting Pt: 2000 Density: 7.13 g/mL |
MDL Number:
MFCD00010933 CAS Number: 1306-38-3 EINECS: 215-150-4 Merck Index: 12,02035 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...