Cobalt(II) sulfamate hydrate

Supplier: BeanTown Chemical

135305-100G 135305-25G 135305-500G
BT135305-100GEA 107 USD
BT135305-100G BT135305-25G BT135305-500G
Cobalt(II) sulfamate hydrate
Cobalt(II) sulfamate hydrate

CAS: 14017-41-5; EC No: 237-834-1; MDL No: MFCD00049877 Powder; Linear Formula: Co(SO3NH2)2·xH2O; MW: 251.11 (anhydrous)


  • Powder

Formula: CoH₄N₂O₆S₂
MDL Number: MFCD00049877
CAS Number: 14017-41-5
EINECS: 237-834-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR