trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl ≥97%
Supplier: BeanTown Chemical
Synonyms:
(trans,trans)-4-(p-Tolyl)-4'-vinyl-1,1'-bi(cyclohexane), trans,trans-4-(p-Tolyl)-4'-vinylbicyclohexyl
135290-1G
135290-5G
BT135290-1GEA
103
USD
BT135290-1G
BT135290-5G
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl ≥97%
trans,trans-4-(4-Methylphenyl)-4'-vinylbicyclohexyl
CAS: 155041-85-3; MDL No: MFCD12911573
Solid; Molecular Formula: C21H30; MW: 282.47
Melting Point: 64-66°
- Solid
Formula:
C₂₁H₃₀ Melting Pt: 64…66 °C |
MDL Number:
MFCD12911573 CAS Number: 155041-85-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...