2,3-Dichloroanisole ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,2-Dichloro-3-methoxybenzene

135280-50G 135280-5G
BT135280-50GEA 138 USD
BT135280-50G BT135280-5G
2,3-Dichloroanisole ≥97%
2,3-Dichloroanisole

CAS: 1984-59-4; EC No: 217-853-1; MDL No: MFCD00000545 Crystalline; Molecular Formula: C7H6Cl2O; MW: 177.03 Melting Point: 31-33°; Flash point: 110°C (230°F)


  • Crystalline

Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Melting Pt: 31…33 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000545
CAS Number: 1984-59-4
EINECS: 217-853-1

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR