ω-Chloro-ω,ω-difluoroacetophenone ≥96%
Supplier: BeanTown Chemical
135265-1G
135265-5G
BT135265-1GEA
34
USD
BT135265-1G
BT135265-5G
ω-Chloro-ω,ω-difluoroacetophenone ≥96%
ω-Chloro-ω,ω-difluoroacetophenone
CAS: 384-67-8; MDL No: MFCD00042123
Liquid; Molecular Formula: C8H5ClF2O; MW: 190.58
Flash point: 72°C (161°F)
Refractive Index: 1.497
- Refractive index: 1.497
- Liquid
Formula:
(C₆H₅)C(O)CClF₂ Flash Pt: 72 °C (161 °F) |
MDL Number:
MFCD00042123 CAS Number: 384-67-8 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...