(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide ≥98%
Supplier: BeanTown Chemical
135260-1G
135260-250MG
135260-5G
BT135260-1GEA
98
USD
BT135260-1G
BT135260-250MG
BT135260-5G
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide ≥98%
(S)-N-tert-Butyldecahydroisoquinoline-3-carboxamide
CAS: 136465-81-1; EC No: 420-380-5; MDL No: MFCD01313226
Solid; Molecular Formula: C14H26N2O ; MW: 238.37
Melting Point: 112-115°
Optical Rotation: [α]22/D -71°, c = 1 in methanol
- Optical rotation: [α]22/D -71°, c = 1 in methanol
- Solid
Formula:
C₁₄H₂₆N₂O Melting Pt: 112…115 °C |
MDL Number:
MFCD01313226 CAS Number: 136465-81-1 EINECS: 420-380-5 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...