4,5-Dibromo-1H-1,2,3-triazole

Supplier: BeanTown Chemical

135255-1G 135255-5G
BT135255-1GEA 112 USD
BT135255-1G BT135255-5G
4,5-Dibromo-1H-1,2,3-triazole
4,5-Dibromo-1H-1,2,3-triazole

CAS: 15294-81-2 Solid; Molecular Formula: C2HBr2N3; MW: 226.86 Boiling Point: 348° Density (g/mL): 2.62


  • Solid

Formula: C₂HBr₂N₃
MW: 226.859 g/mol
Boiling Pt: 348 °C
Density: 2.62 g/mL
MDL Number: MFCD08062401
CAS Number: 15294-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR