Methyl-4-iodobenzoate ≥98%
Supplier: BeanTown Chemical
135225-10G
135225-50G
BT135225-10GEA
46
USD
BT135225-10G
BT135225-50G
Methyl-4-iodobenzoate ≥98%
Methyl-4-iodobenzoate
CAS: 619-44-3; EC No: 210-597-1; MDL No: MFCD00016353
Solid; Molecular Formula: C8H7IO2; MW: 262.05
Melting Point: 112-116°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₈H₇IO₂ Melting Pt: 112…116 °C |
MDL Number:
MFCD00016353 CAS Number: 619-44-3 EINECS: 210-597-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...