2-Chloro-4-iodo-3-methylpyridine ≥95%
Supplier: BeanTown Chemical
135150-1G
135150-5G
BT135150-1GEA
205
USD
BT135150-1G
BT135150-5G
2-Chloro-4-iodo-3-methylpyridine ≥95%
2-Chloro-4-iodo-3-methylpyridine
CAS: 153034-88-9; MDL No: MFCD03095293
Solid; Molecular Formula: C6H5ClIN; MW: 253.47
Melting Point: 101-106°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₆H₅ClIN Melting Pt: 101…106 °C |
MDL Number:
MFCD03095293 CAS Number: 153034-88-9 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...