Copper ≥99.9% (trace metals basis), shot 1-10 mm

Supplier: BeanTown Chemical

135145-2KG 135145-500G
BT135145-2KGEA 136.65 USD
BT135145-2KG BT135145-500G
Copper ≥99.9% (trace metals basis), shot 1-10 mm
Copper

  • RTECS: GL5325000
  • Solid

Formula: Cu
Boiling Pt: 2567 °C
Melting Pt: 1083.4 °C
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR