Potassium disulfite ≥95%
Supplier: BeanTown Chemical
|
Warning
|
Synonyms:
Dipotassium disulphite, Potassium pyrosulphite, Potassium metabisulphite, di-Potassium disulphite
135070-100G
135070-2KG
135070-500G
BT135070-100GEA
40.89
USD
BT135070-100G
BT135070-2KG
BT135070-500G
Potassium disulfite ≥95%
Potassium disulphite
CAS: 16731-55-8; EC No: 240-795-3; MDL No: MFCD00167605
Powder; Linear Formula: K2S2O5; MW: 222.32
Melting Point: 190° (decomposes)
Density (g/mL): 2.34
Air Sensitive
Formula:
K₂O₅S₂ MW: 222.33 g/mol Melting Pt: 190 °C (dec.) Density: 2,34 g/mL |
MDL Number:
MFCD00167605 CAS Number: 16731-55-8 EINECS: 240-795-3 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...