1,8-Naphthalenedicarboxylic anhydride ≥97%

Supplier: BeanTown Chemical

Synonyms: Naphthalene-1,8-dicarboxylic anhydride, Benzo[de]isochromene-1,3-dione, 1,8-Naphthalic anhydride

135065-1KG 135065-250G 135065-50G
BT135065-1KGEA 152 USD
BT135065-1KG BT135065-250G BT135065-50G
1,8-Naphthalenedicarboxylic anhydride ≥97%
1,8-Naphthalenedicarboxylic anhydride

CAS: 81-84-5; EC No: 201-380-2; MDL No: MFCD00006925; RTECS: QK5350000 Powder; Molecular Formula: C12H6O3 ; MW: 198.17 Melting Point: 267-269°; Flash point: 272°C (522°F) Moisture Sensitive


  • RTECS: QK5350000
  • Moisture Sensitive
  • Powder

Formula: C₁₂H₆O₃
MW: 198.18 g/mol
Melting Pt: 267…269 °C
Flash Pt: 272 °C (522 °F)
MDL Number: MFCD00006925
CAS Number: 81-84-5
EINECS: 201-380-2

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR