1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate ≥97%

Supplier: BeanTown Chemical

Synonyms: Catechol-3,5-disulphonic acid disodium salt monohydrate, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt monohydrate, Tiron monohydrate, 3,5-Pyrocatecholdisulphonic acid disodium salt monohydrate, Disodium 4,5-dihydroxybenzene-1,3-disulphonate monohydrate

135055-100G 135055-25G 135055-500G
BT135055-100GEA 83 USD
BT135055-100G BT135055-25G BT135055-500G
1,2-Dihydroxybenzene-3,5-disulfonic acid disodium salt monohydrate ≥97%
1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt monohydrate

CAS: 270573-71-2; EC No: 205-741-5; MDL No: MFCD00007473; RTECS: CZ9263000 Solid; Molecular Formula: C6H4Na2O8S2·H2O; MW: 332.22 Melting Point: 300° Hygroscopic


  • RTECS: CZ9263000
  • Hygroscopic
  • Solid

Formula: C₆H₄Na₂O₈S₂·1H₂O
MW: 332.22 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00149531
CAS Number: 270573-71-2
EINECS: 205-741-5

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR