2-Bromo-1-tetralone
Supplier: BeanTown Chemical
Synonyms:
2-Bromo-3,4-dihydronaphthalen-1(2H)-one, 2-Bromo-3,4-dihydro-1(2H)-naphthalenone
135035-1G
135035-5G
BT135035-1GEA
135
USD
BT135035-1G
BT135035-5G
2-Bromo-1-tetralone
2-Bromo-1-tetralone
CAS: 13672-07-6; EC No: 237-154-5; MDL No: MFCD00089999
Solid; Molecular Formula: C10H9BrO; MW: 225.08
Melting Point: 39-43°
- Solid
Formula:
C₁₀H₉BrO MW: 225.08 g/mol Melting Pt: 39…43 °C |
MDL Number:
MFCD00089999 CAS Number: 13672-07-6 EINECS: 237-154-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...