2-Chloro-6-methylpyrazine ≥98%

Supplier: BeanTown Chemical

135025-1G 135025-250MG 135025-5G
BT135025-1GEA 95 USD
BT135025-1G BT135025-250MG BT135025-5G
2-Chloro-6-methylpyrazine ≥98%
2-Chloro-6-methylpyrazine

CAS: 38557-71-0; MDL No: MFCD00055032 Solid; Molecular Formula: C5H5ClN2 ; MW: 128.56


  • Solid

Formula: C₅H₅ClN₂
MW: 128.5596 g/mol
MDL Number: MFCD00055032
CAS Number: 38557-71-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR