Calcium sulfate, anhydrous ≥99%

Supplier: BeanTown Chemical

134990-1KG 134990-250G 134990-5KG
BT134990-1KGEA 85.27 USD
BT134990-1KG BT134990-250G BT134990-5KG
Calcium sulfate, anhydrous ≥99%
Calcium sulphate

CAS: 7778-18-9; EC No: 231-900-3; MDL No: MFCD00010912; RTECS: WS6920000 Powder; Linear Formula: CaSO4; MW: 136.14 Hygroscopic


  • RTECS: WS6920000
  • Hygroscopic
  • Powder

Formula: CaO₄S
MDL Number: MFCD00010912
CAS Number: 7778-18-9
EINECS: 231-900-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR