Palladium silver alloy (75:25 w%) ≥99.9% (trace metals basis excluding Pt), foil, Thickness 0.5 mm
Supplier: BeanTown Chemical
134975-25X25MM
134975-50X50MM
BT134975-25X25EA
776
USD
BT134975-25X25
BT134975-50X50
Palladium silver alloy (75:25 w%) ≥99.9% (trace metals basis excluding Pt), foil, Thickness 0.5 mm
Palladium silver alloy (75:25 w%)
MDL No: MFCD02091738
Solid; Formula: Pd:Ag; 75:25 wt%
- Solid
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...