Ammonium dihydrogen phosphate ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

134965-1G 134965-5G
BT134965-1GEA 79 USD
BT134965-1G BT134965-5G
Ammonium dihydrogen phosphate ≥99.995%
Ammonium dihydrogen phosphate

CAS: 7722-76-1; EC No: 231-764-5; MDL No: MFCD00003396; RTECS: TC6587000 Crystalline; Linear Formula: NH4H2PO4; MW: 115.03 Melting Point: 190° (decomposes)


  • RTECS: TC6587000
  • Crystalline

Formula: NH₄H₂PO₄
Melting Pt: 190 °C (dec.)
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR