2-Amino-6-bromobenzothiazole ≥98%

Supplier: BeanTown Chemical

134955-1G 134955-25G 134955-5G
BT134955-1GEA 57 USD
BT134955-1G BT134955-25G BT134955-5G
2-Amino-6-bromobenzothiazole ≥98%
2-Amino-6-bromobenzothiazole

CAS: 15864-32-1; MDL No: MFCD00152229 Crystalline/Powder; Molecular Formula: C7H5BrN2S; MW: 229.10 Melting Point: 213-217°


  • Crystalline/Powder

Formula: C₇H₅BrN₂S
MW: 229.1
Melting Pt: 213…217 °C
MDL Number: MFCD00152229
CAS Number: 15864-32-1

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR