Barbituric acid ≥99%
Supplier: BeanTown Chemical
134930-100G
134930-25G
134930-500G
BT134930-100GEA
48.53
USD
BT134930-100G
BT134930-25G
BT134930-500G
Barbituric acid ≥99%
Barbituric acid
CAS: 67-52-7; EC No: 200-658-0; MDL No: MFCD00006666; RTECS: CP8000000
Solid; Molecular Formula: C4H4N2O3; MW: 128.09
Melting Point: 248-252° (decomposes); Flash point: 150°C (302°F)
- RTECS: CP8000000
- Solid
Formula:
C₄H₄N₂O₃ Melting Pt: 248…252 °C (dec.) Flash Pt: 150 °C (302 °F) |
MDL Number:
MFCD00006666 CAS Number: 67-52-7 EINECS: 200-658-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...