tert-Butyl-2-((diphenylmethylene)amino)acetate ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-(Diphenylmethylene)glycine tert-butyl ester, Glycine, N-(diphenylmethylene)-, 1,1-dimethylethyl ester, tert-Butyl 2-((diphenylmethylene)amino)acetate
134725-1G
134725-5G
BT134725-1GEA
85
USD
BT134725-1G
BT134725-5G
tert-Butyl-2-((diphenylmethylene)amino)acetate ≥99%
tert-Butyl-2-((diphenylmethylene)amino)acetate
CAS: 81477-94-3; MDL No: MFCD00134280
Powder; Linear Formula: (C6H5)2C=NCH2COOC(CH3)3; Molecular Formula: C19H21NO2 ; MW: 295.38
Melting Point: 114-115°
- Powder
Formula:
C₁₉H₂₁NO₂ Melting Pt: 114…115 °C |
MDL Number:
MFCD00134280 CAS Number: 81477-94-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...