4,6-Dihydroxypyrimidine ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Hydroxy-1H-pyrimidin-4-one
134720-100G
134720-25G
134720-500G
BT134720-100GEA
138
USD
BT134720-100G
BT134720-25G
BT134720-500G
4,6-Dihydroxypyrimidine ≥98%
4,6-Dihydroxypyrimidine
CAS: 1193-24-4; EC No: 214-772-3; MDL No: MFCD00016733; RTECS: UW7523000
Powder; Molecular Formula: C4H4N2O2; MW: 112.09
Melting Point:
- RTECS: UW7523000
- Powder
Formula:
C₄H₄N₂O₂ MW: 112.09 g/mol Melting Pt: >300 °C |
MDL Number:
MFCD00016733 CAS Number: 1193-24-4 EINECS: 214-772-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...