Methyl-5-bromo-2-furoate ≥97%
Supplier: BeanTown Chemical
Synonyms:
5-Bromo-2-furoic acid methyll ester, 5-Bromo-2-furancarboxylic acid methyll ester, Methyl 5-Bromo-2-furancarboxylate, Methyl 5-Bromo-2-furoate, Methyl 5-bromofuran-2-carboxylate
134705-1G
134705-5G
BT134705-1GEA
65
USD
BT134705-1G
BT134705-5G
Methyl-5-bromo-2-furoate ≥97%
Methyl-5-bromo-2-furoate
CAS: 2527-99-3; MDL No: MFCD00092313
Solid; Molecular Formula: C6H5BrO3; MW: 205.01
Melting Point: 62-65°
- Solid
Formula:
C₆H₅BrO₃ MW: 205.01 g/mol Melting Pt: 62…65 °C |
MDL Number:
MFCD00092313 CAS Number: 2527-99-3 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...