2-Methyl-5-nitroimidazole-1-ethanol ≥99%
Supplier: BeanTown Chemical
Synonyms:
Metronidazole
134695-100G
134695-25G
BT134695-100GEA
228
USD
BT134695-100G
BT134695-25G
2-Methyl-5-nitroimidazole-1-ethanol ≥99%
2-Methyl-5-nitroimidazole-1-ethanol
CAS: 443-48-1; EC No: 207-136-1; MDL No: MFCD00009750; RTECS: NI5600000
Crystalline/Powder; Molecular Formula: C6H9N2O3; MW: 171.15
Melting Point: 159-161°
- RTECS: NI5600000
- Crystalline/Powder
Formula:
C₆H₉N₃O₃ Melting Pt: 159…161 °C |
MDL Number:
MFCD00009750 CAS Number: 443-48-1 EINECS: 207-136-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...