Silicon nitride ≥98.5%
Supplier: BeanTown Chemical
Synonyms:
Silicon(IV) nitride
134690-100G
134690-25G
134690-5G
BT134690-100GEA
495
USD
BT134690-100G
BT134690-25G
BT134690-5G
Silicon nitride ≥98.5%
Silicon nitride
CAS: 12033-89-5; EC No: 234-796-8; MDL No: MFCD00011230; RTECS: VW0650000
15-30nm APS Powder, S.A. 103-123 m2/g; Linear Formula: Si3N4; MW: 140.28
Melting Point: 1900° (sublimes)
Density (g/mL): 3.44
- RTECS: VW0650000
- 15-30nm APS Powder, S.A. 103-123 m²/g
Formula:
N₄Si₃ MW: 140.28 g/mol Melting Pt: 1900 °C (subl.) Density: 3.44 g/mL |
MDL Number:
MFCD00011230 CAS Number: 12033-89-5 EINECS: 234-796-8 |
Specification Test Results
Assay | 98.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...