3-Amino-3-(2-nitrophenyl)propionic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Amino-3-(2-nitrophenyl)propanoic acid

134685-100G 134685-25G 134685-5G
BT134685-100GEA 976 USD
BT134685-100G BT134685-25G BT134685-5G
3-Amino-3-(2-nitrophenyl)propionic acid ≥98%
3-Amino-3-(2-nitrophenyl)propionic acid

CAS: 5678-48-8; MDL No: MFCD00090356 Powder; Molecular Formula: C9H10N2O4 ; MW: 210.19


  • Powder

Formula: C₉H₁₀N₂O₄
MW: 210.19 g/mol
MDL Number: MFCD00090356
CAS Number: 5678-48-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR