N,N'-Di(1-naphthyl)benzidine ≥98%

Supplier: BeanTown Chemical

Synonyms: N,N’-Di(1-naphthyl)-4,4’-benzidine

134680-1G 134680-250MG 134680-5G
BT134680-1GEA 70 USD
BT134680-1G BT134680-250MG BT134680-5G
N,N'-Di(1-naphthyl)benzidine ≥98%
N,N'-Di(1-naphthyl)benzidine

CAS: 152670-41-2; MDL No: MFCD09261380 Solid; Molecular Formula: C32H24N2; MW: 436.56 Melting Point: 252-258° Refractive Index: 1.229


  • Refractive index: 1.229
  • Solid

Formula: C₃₂H₂₄N₂
MW: 436.558 g/mol
Melting Pt: 252…258 °C
MDL Number: MFCD09261380
CAS Number: 152670-41-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR