2-Methoxy-5-pyridineboronic acid ≥95%

Supplier: BeanTown Chemical

Synonyms: (6-Methoxypyridin-3-yl)boronic acid

134655-1G 134655-250MG 134655-5G
BT134655-1GEA 262 USD
BT134655-1G BT134655-250MG BT134655-5G
2-Methoxy-5-pyridineboronic acid ≥95%
2-Methoxy-5-pyridineboronic acid

CAS: 163105-89-3; MDL No: MFCD02093044 Solid; Molecular Formula: C6H8BNO3; MW: 152.94 Melting Point: 135-140°


  • Solid

Formula: C₆H₈BNO₃
MW: 152.95 g/mol
Melting Pt: 135…140 °C
MDL Number: MFCD02093044
CAS Number: 163105-89-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR