Methyl-2-amino-4-chlorobenzoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-amino-4-chlorobenzoate

134625-10G 134625-50G
BT134625-10GEA 82 USD
BT134625-10G BT134625-50G
Methyl-2-amino-4-chlorobenzoate ≥98%
Methyl-2-amino-4-chlorobenzoate

CAS: 5900-58-3; EC No: 227-593-0; MDL No: MFCD00017568 Crystalline; Molecular Formula: C8H8ClNO2; MW: 185.61 Melting Point: 66-71°


  • Crystalline

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 66…71 °C
MDL Number: MFCD00017568
CAS Number: 5900-58-3
EINECS: 227-593-0

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR