Diethyl methylphosphonate 97
Supplier: BeanTown Chemical
134570-10G
134570-50G
BT134570-10GEA
119.46
USD
BT134570-10G
BT134570-50G
Diethyl methylphosphonate 97
Diethyl methylphosphonate
CAS: 683-08-9; EC No: 211-667-4; MDL No: MFCD00009813; RTECS: SZ9085000
Liquid; Molecular Formula: C5H13O3P; MW: 152.13
Boiling Point: 194°; Flash point: 75°C (167°F)
Density (g/mL): 1.041; Refractive Index: 1.414
- RTECS: SZ9085000
- Refractive index: 1.414
- Liquid
Formula:
C₅H₁₃O₃P MW: 152.13 g/mol Boiling Pt: 194 °C Density: 1.041 g/mL Flash Pt: 75 °C (167 °F) |
MDL Number:
MFCD00009813 CAS Number: 683-08-9 EINECS: 211-667-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...