Magnesium sulfate hydrate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

134560-10G 134560-50G
BT134560-10GEA 84.95 USD
BT134560-10G BT134560-50G
Magnesium sulfate hydrate ≥99.995% (trace metals basis)
Magnesium sulphate hydrate

CAS: 22189-08-8; EC No: 231-298-2; MDL No: MFCD00149787; RTECS: OM4500000 Solid; Linear Formula: MgSO4·xH2O; MW: 120.37 (anhydrous) Density (g/mL): 1.68 Hygroscopic


  • RTECS: OM4500000
  • Hygroscopic
  • Solid

Formula: MgSO₄·nH₂O
Density: 1.68 g/mL
MDL Number: MFCD00011110
CAS Number: 22189-08-8
EINECS: 231-298-2

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR